accessibility menu, dialog, popup

UserName

CAS RN 41459-42-1

OOBrBr

CAS RN 41459-42-1

IUPAC Name: 3-bromo-2-(bromomethyl)propanoate
Synonyms: 3-bromo-2-(bromomethyl)propanoate
Molecular Weight (g/mol): 244.89
Molecular Formula: C4H5Br2O2
InChi Key: QQZJWQCLWOQDQV-UHFFFAOYSA-M
SMILES: [O-]C(=O)C(CBr)CBr
Quantity 
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results