accessibility menu, dialog, popup

UserName

CAS RN 417721-69-8

NClIH2N

CAS RN 417721-69-8

IUPAC Name: 4-chloro-3-iodopyridin-2-amine
Synonyms: 4-chloro-3-iodopyridin-2-amine
Molecular Weight (g/mol): 254.46
Molecular Formula: C5H4ClIN2
InChi Key: BWMULFKDCVPGNF-UHFFFAOYSA-N
SMILES: NC1=NC=CC(Cl)=C1I

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results