accessibility menu, dialog, popup

UserName

H3CCH3OOOH

CAS RN 4191-73-5

IUPAC Name: propan-2-yl 4-hydroxybenzoate
Synonyms: isopropylparaben
Molecular Weight (g/mol): 180.2
Molecular Formula: C10H12O3
InChi Key: CMHMMKSPYOOVGI-UHFFFAOYSA-N
SMILES: CC(C)OC(=O)C1=CC=C(O)C=C1
Molecular Weight (g/mol) 
  • (3)
  • (4)
Percent Purity 
  • (3)
  • (4)
Quantity 
  • (1)
  • (2)
  • (1)
  • (2)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (3)
  • (3)
  • (4)
  • (3)
  • (4)
  • (1)
  • (2)
  • (1)
  • (2)
  • (1)