accessibility menu, dialog, popup

UserName

CAS RN 4197-07-3

NaNaOHNHNOSOOOSOOO(Z)

CAS RN 4197-07-3

IUPAC Name: disodium (3Z)-5-hydroxy-4-oxo-3-(2-phenylhydrazin-1-ylidene)-3,4-dihydronaphthalene-2,7-disulfonate
Synonyms: disodium (3Z)-5-hydroxy-4-oxo-3-(2-phenylhydrazin-1-ylidene)naphthalene-2,7-disulfonate
Molecular Weight (g/mol): 468.36
Molecular Formula: C16H10N2Na2O8S2
InChi Key: XTJONEUTTZZRAB-VZRGYXGKSA-L
SMILES: [Na+].[Na+].OC1=CC(=CC2=C1C(=O)\C(=N\NC1=CC=CC=C1)C(=C2)S([O-])(=O)=O)S([O-])(=O)=O
Quantity 
  • (2)
  • (2)
Color 
  • (2)
Grade 
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (2)