accessibility menu, dialog, popup

UserName

HClCH3NH3CO

CAS RN 42036-65-7

IUPAC Name: hydrogen 2-[(dimethylamino)methyl]cyclohexan-1-one chloride
Synonyms: hydrogen 2-[(dimethylamino)methyl]cyclohexan-1-one chloride
Molecular Weight (g/mol): 191.7
Molecular Formula: C9H18ClNO
InChi Key: CLVHTSWMNNSUSH-UHFFFAOYNA-N
SMILES: [H+].[Cl-].CN(C)CC1CCCCC1=O
Quantity 
  • (1)
  • (1)
  • (1)
  • (1)
Physical Form 
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results