CAS RN 420841-84-5
IUPAC Name:
2-[3-(4-nitrophenoxy)phenyl]-1,3-dioxo-2,3-dihydro-1H-isoindole-5-carboxylic acid
Synonyms:
2-[3-(4-nitrophenoxy)phenyl]-1,3-dioxoisoindole-5-carboxylic acid
Molecular Weight (g/mol):
404.33
Molecular Formula:
C21H12N2O7
InChi Key:
HFYPTENHTPNXGP-UHFFFAOYSA-N
SMILES:
OC(=O)C1=CC2=C(C=C1)C(=O)N(C2=O)C1=CC(OC2=CC=C(C=C2)[N+]([O-])=O)=CC=C1
1
–
1
of
1
results
1
H2L 5765834, R&D Systems™
CAS: 420841-84-5 Molecular Formula: C21H12N2O7 Molecular Weight (g/mol): 404.334 InChI Key: HFYPTENHTPNXGP-UHFFFAOYSA-N Synonym: 2-3-4-nitrophenoxy phenyl-1,3-dioxoisoindole-5-carboxylic acid,2-3-4-nitrophenoxy phenyl-1,3-dioxoisoindoline-5-carboxylic acid,2,3-dihydro-2-3-4-nitrophenoxy phenyl-1,3-dioxo-1h-isoindole-5-carboxylic acid,2-3-4-nitrophenoxy phenyl-1,3-dioxo-2,3-dihydro-1h-isoindole-5-carboxylic acid PubChem CID: 1365686 IUPAC Name: 2-[3-(4-nitrophenoxy)phenyl]-1,3-dioxoisoindole-5-carboxylic acid SMILES: C1=CC(=CC(=C1)OC2=CC=C(C=C2)[N+](=O)[O-])N3C(=O)C4=C(C3=O)C=C(C=C4)C(=O)O