accessibility menu, dialog, popup

UserName

CAS RN 42303-42-4

CH3OONH2HCl

CAS RN 42303-42-4

IUPAC Name: ethyl 1-aminocyclopropane-1-carboxylate hydrochloride
Synonyms: ethyl 1-aminocyclopropane-1-carboxylate hydrochloride
Molecular Weight (g/mol): 165.62
Molecular Formula: C6H12ClNO2
InChi Key: XFNUTZWASODOQK-UHFFFAOYSA-N
SMILES: Cl.CCOC(=O)C1(N)CC1
Physical Form 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (2)
  • (1)
  • (1)