accessibility menu, dialog, popup

UserName

CAS RN 425-61-6

OHFFFFHO

CAS RN 425-61-6

IUPAC Name: 2,2,3,3-tetrafluorobutane-1,4-diol
Synonyms: 2,2,3,3-tetrafluorobutane-1,4-diol
Molecular Weight (g/mol): 162.08
Molecular Formula: C4H6F4O2
InChi Key: CDZXJJOGDCLNKX-UHFFFAOYSA-N
SMILES: OCC(F)(F)C(F)(F)CO
Boiling Point 
  • (3)
Quantity 
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (3)
  • (3)
  • (1)
  • (1)
  • (1)