accessibility menu, dialog, popup

UserName

CH3H2CBOHOH

CAS RN 42599-18-8

IUPAC Name: (hex-5-en-2-yl)boronic acid
Synonyms: hex-5-en-2-ylboronic acid
Molecular Weight (g/mol): 127.98
Molecular Formula: C6H13BO2
InChi Key: HLAPIUQASQCHKW-UHFFFAOYNA-N
SMILES: CC(CCC=C)B(O)O

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results