accessibility menu, dialog, popup

UserName

CAS RN 4274-38-8

HClNH2HSFFF

CAS RN 4274-38-8

IUPAC Name: hydrogen 2-amino-4-(trifluoromethyl)benzene-1-thiol chloride
Synonyms: hydrogen 2-amino-4-(trifluoromethyl)benzenethiol chloride
Molecular Weight (g/mol): 229.65
Molecular Formula: C7H7ClF3NS
InChi Key: FIAGYDIJZOWVAB-UHFFFAOYSA-N
SMILES: [H+].[Cl-].NC1=CC(=CC=C1S)C(F)(F)F
Quantity 
  • (2)
  • (1)
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results