CAS RN 4318-76-7
IUPAC Name:
pyridine-2,5-diamine
Synonyms:
2,5-diaminopyridine
Molecular Weight (g/mol):
109.13
Molecular Formula:
C5H7N3
InChi Key:
MIROPXUFDXCYLG-UHFFFAOYSA-N
SMILES:
NC1=CC=C(N)N=C1
1
–
1
of
1
results
1
2,5-Diaminopyridine, 97%
CAS: 4318-76-7 Molecular Formula: C5H7N3 Molecular Weight (g/mol): 109.13 InChI Key: MIROPXUFDXCYLG-UHFFFAOYSA-N Synonym: 2,5-diaminopyridine,2,5-pyridinediamine,pyridine, 2,5-diamino,diaminopyridine van,unii-h6olz060hi,h6olz060hi,da1,pyridine,5-diamino,2,5-diamino pyridine,2,5-diamino-pyridine PubChem CID: 20314 IUPAC Name: pyridine-2,5-diamine SMILES: C1=CC(=NC=C1N)N