CAS RN 43197-78-0
IUPAC Name:
3-chloro-3-methylhexane
Synonyms:
3-chloro-3-methylhexane
Molecular Weight (g/mol):
134.65
Molecular Formula:
C7H15Cl
InChi Key:
UTKDCNDVTOWUHW-UHFFFAOYNA-N
SMILES:
CCCC(C)(Cl)CC
1
–
1
of
1
results
1
3-Chloro-3-methylhexane, 97%, Thermo Scientific™
CAS: 43197-78-0 Molecular Formula: C7H15Cl Molecular Weight (g/mol): 134.65 MDL Number: MFCD00060778 InChI Key: UTKDCNDVTOWUHW-UHFFFAOYNA-N Synonym: hexane, 3-chloro-3-methyl,acmc-20an6z,3-chlor-3-methyl-hexan,hexane,3-chloro-3-methyl PubChem CID: 142659 IUPAC Name: 3-chloro-3-methylhexane SMILES: CCCC(C)(Cl)CC