accessibility menu, dialog, popup

UserName

FFFFFFFFFF(?)

CAS RN 434-90-2

IUPAC Name: decafluoro-1,1'-biphenyl
Synonyms: decafluorobiphenyl
Molecular Weight (g/mol): 334.12
Molecular Formula: C12F10
InChi Key: ONUFSRWQCKNVSL-UHFFFAOYSA-N
SMILES: FC1=C(F)C(F)=C(C(F)=C1F)C1=C(F)C(F)=C(F)C(F)=C1F
Molecular Weight (g/mol) 
  • (1)
  • (9)
  • (2)
Solvent or Matrix 
  • (1)
  • (2)
  • (1)
  • (2)
  • (1)
Quantity 
  • (1)
  • (1)
  • (1)
  • (2)
Boiling Point 
  • (2)
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (3)
  • (4)
  • (2)
  • (2)
  • (1)
  • (1)
  • (9)
  • (2)
  • (1)
  • (2)
  • (1)
  • (2)
  • (1)
  • (1)
  • (1)
  • (1)
  • (2)
  • (2)
  • (2)
  • (3)
  • (2)