accessibility menu, dialog, popup

UserName

Br(E)

CAS RN 4392-24-9

IUPAC Name: [(1E)-3-bromoprop-1-en-1-yl]benzene
Synonyms: (3-bromo-propenyl)-benzene
Molecular Weight (g/mol): 197.08
Molecular Formula: C9H9Br
InChi Key: RUROFEVDCUGKHD-QPJJXVBHSA-N
SMILES: BrC\C=C\C1=CC=CC=C1
Molecular Weight (g/mol) 
  • (8)
  • (3)
Quantity 
  • (1)
  • (2)
  • (1)
  • (3)
  • (1)
  • (3)
Percent Purity 
  • (3)
  • (3)
  • (2)
Boiling Point 
  • (3)
  • (3)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (3)
  • (3)
  • (3)
  • (2)
  • (8)
  • (3)
  • (1)
  • (2)
  • (1)
  • (3)
  • (1)
  • (3)
  • (3)
  • (3)
  • (2)
  • (3)
  • (3)
  • (3)