CAS RN 440124-96-9
IUPAC Name:
—
Synonyms:
—
Molecular Weight (g/mol):
—
Molecular Formula:
—
InChi Key:
—
SMILES:
—
1
–
1
of
1
results
1
(E)-Dihydro Efavirenz, TRC
CAS: 440124-96-9 Molecular Formula: C14 H11 Cl F3 N O2 Molecular Weight (g/mol): 317.69 Synonym: (4S)-6-Chloro-4-[(1E)-2-cyclopropylethenyl]-1,4-dihydro-4-(trifluoromethyl)-2H-3,1-benzoxazin-2-one,SR 695,(S,E)-6-Chloro-4-(2-cyclopropylvinyl)-4-(trifluoromethyl)-2H-3,1-benzoxazin-2-one,Efavirenz Related Compound B (USP) IUPAC Name: (4S)-6-chloro-4-[(E)-2-cyclopropylethenyl]-4-(trifluoromethyl)-1H-3,1-benzoxazin-2-one SMILES: FC(F)(F)[C@]1(OC(=O)Nc2ccc(Cl)cc12)\C=C\C3CC3