accessibility menu, dialog, popup

UserName

CAS RN 4426-83-9

H3CNCS

CAS RN 4426-83-9

IUPAC Name: 1-isothiocyanatoheptane
Synonyms: 1-isothiocyanatoheptane
Molecular Weight (g/mol): 157.28
Molecular Formula: C8H15NS
InChi Key: LIPUQNPCPLDDBO-UHFFFAOYSA-N
SMILES: CCCCCCCN=C=S

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results