CAS RN 444-27-9
CAS RN 444-27-9
IUPAC Name:
—
Synonyms:
—
Molecular Weight (g/mol):
—
Molecular Formula:
—
InChi Key:
—
SMILES:
—
1
–
1
of
1
results
1
Timonacic, MedChemExpress
MedChemExpress Timonacic is used as an adjuvant in the treatment of acute and hepatic disorders. It has also been used for the treatment of some cases of cancer, through the induction of the reverse transformation.