accessibility menu, dialog, popup

UserName

CAS RN 4448-77-5

H2N

CAS RN 4448-77-5

IUPAC Name: 1-cycloheptylmethanamine
Synonyms: 1-cycloheptylmethanamine
Molecular Weight (g/mol): 127.23
Molecular Formula: C8H17N
InChi Key: CAOQEOHEZKVYOJ-UHFFFAOYSA-N
SMILES: NCC1CCCCCC1
Boiling Point 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results