accessibility menu, dialog, popup

UserName

CAS RN 4491-92-3

IUPAC Name: 3-(4-methylpiperazin-1-yl)propanenitrile
Synonyms: 3-(4-methylpiperazin-1-yl)propanenitrile
Molecular Weight (g/mol): 153.23
Molecular Formula: C8H15N3
InChi Key: XMZMOGFGIJQCHN-UHFFFAOYSA-N
SMILES: CN1CCN(CCC#N)CC1
Boiling Point 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results