CAS RN 4494-53-5
IUPAC Name:
—
Synonyms:
—
Molecular Weight (g/mol):
—
Molecular Formula:
—
InChi Key:
—
SMILES:
—
1
–
1
of
1
results
1
2-Methyl-1,3(2H,4H)-isoquinolinedione, TRC
CAS: 4494-53-5 Molecular Formula: C10H9NO2 Molecular Weight (g/mol): 175.18 Synonym: 2-Methylisoquinoline-1,3(2H,4H)-dione IUPAC Name: 2-methyl-4H-isoquinoline-1,3-dione SMILES: CN1C(=O)Cc2ccccc2C1=O