accessibility menu, dialog, popup

UserName

CAS RN 450-93-1

H3COFHO

CAS RN 450-93-1

IUPAC Name: 4-fluoro-2-methoxyphenol
Synonyms: 4-fluoro-2-methoxyphenol
Molecular Weight (g/mol): 142.13
Molecular Formula: C7H7FO2
InChi Key: OULGLTLTWBZBLO-UHFFFAOYSA-N
SMILES: COC1=CC(F)=CC=C1O
Boiling Point 
  • (3)
  • (2)
Quantity 
  • (2)
  • (1)
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (3)
  • (3)
  • (2)
  • (2)
  • (1)
  • (2)
  • (2)