accessibility menu, dialog, popup

UserName

ClO

CAS RN 4524-93-0

IUPAC Name: cyclopentanecarbonyl chloride
Synonyms: cyclopentanecarbonyl chloride
Molecular Weight (g/mol): 132.59
Molecular Formula: C6H9ClO
InChi Key: WEPUZBYKXNKSDH-UHFFFAOYSA-N
SMILES: ClC(=O)C1CCCC1
Molecular Weight (g/mol) 
  • (3)
  • (6)
Quantity 
  • (2)
  • (1)
  • (2)
  • (2)
  • (1)
  • (1)
Boiling Point 
  • (3)
  • (3)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (3)
  • (3)
  • (6)
  • (2)
  • (1)
  • (2)
  • (2)
  • (1)
  • (1)
  • (3)
  • (3)
  • (3)