accessibility menu, dialog, popup

UserName

CAS RN 4543-96-8

H3CNH2NHCH3CH3

CAS RN 4543-96-8

IUPAC Name: [3-(dimethylazaniumyl)propyl](methyl)azanium
Synonyms: [3-(dimethylammonio)propyl](methyl)azanium
Molecular Weight (g/mol): 118.22
Molecular Formula: C6H18N2
InChi Key: SORARJZLMNRBAQ-UHFFFAOYSA-P
SMILES: C[NH2+]CCC[NH+](C)C
Quantity 
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results