accessibility menu, dialog, popup

UserName

CAS RN 4572-03-6

CH3NNNH2

CAS RN 4572-03-6

IUPAC Name: 3-(4-methylpiperazin-1-yl)propan-1-amine
Synonyms: 3-(4-methylpiperazin-1-yl)propan-1-amine
Molecular Weight (g/mol): 157.26
Molecular Formula: C8H19N3
InChi Key: RGUABPVONIGVAT-UHFFFAOYSA-N
SMILES: CN1CCN(CCCN)CC1
Boiling Point 
  • (3)
Quantity 
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (3)
  • (3)
  • (1)
  • (1)
  • (1)