accessibility menu, dialog, popup

UserName

CAS RN 459-59-6

H3CNHF

CAS RN 459-59-6

IUPAC Name: 4-fluoro-N-methylaniline
Synonyms: 4-fluoro-N-methylaniline
Molecular Weight (g/mol): 125.15
Molecular Formula: C7H8FN
InChi Key: VLWRKVBQUANIGI-UHFFFAOYSA-N
SMILES: CNC1=CC=C(F)C=C1
Boiling Point 
  • (3)
Quantity 
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results