accessibility menu, dialog, popup

UserName

CAS RN 4595-59-9

BrNN

CAS RN 4595-59-9

IUPAC Name: 5-bromopyrimidine
Synonyms: 5-bromopyrimidine
Molecular Weight (g/mol): 158.99
Molecular Formula: C4H3BrN2
InChi Key: GYCPLYCTMDTEPU-UHFFFAOYSA-N
SMILES: BrC1=CN=CN=C1
Molecular Weight (g/mol) 
  • (3)
  • (4)
  • (1)
Quantity 
  • (2)
  • (1)
  • (3)
  • (2)
Physical Form 
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (1)
  • (2)
  • (3)
  • (3)
  • (3)
  • (4)
  • (1)
  • (2)
  • (1)
  • (3)
  • (2)
  • (2)