CAS RN 461-87-0
CAS RN 461-87-0
IUPAC Name:
2-fluoro-4-methylpyridine
Synonyms:
2-fluoro-4-methylpyridine
Molecular Weight (g/mol):
111.12
Molecular Formula:
C6H6FN
InChi Key:
ZBFAXMKJADVOGH-UHFFFAOYSA-N
SMILES:
CC1=CC=NC(F)=C1
1
–
2
of
2
results
1
2-Fluoro-4-methylpyridine, 98%
CAS: 461-87-0 Molecular Formula: C6H6FN Molecular Weight (g/mol): 111.119 MDL Number: MFCD00041224 InChI Key: ZBFAXMKJADVOGH-UHFFFAOYSA-N Synonym: 2-fluoro-4-picoline,2-fluoro-4-methyl-pyridine,pyridine, 2-fluoro-4-methyl,2-fluoro-4-methyl pyridine,pubchem1203,acmc-1aevi,4-methyl-2-fluoropyridine,2-fluoro-4-methylpyridine,,ksc494a2j PubChem CID: 2737403 IUPAC Name: 2-fluoro-4-methylpyridine SMILES: CC1=CC(=NC=C1)F