accessibility menu, dialog, popup

UserName

HOBOHBOHHO

CAS RN 4612-26-4

IUPAC Name: [4-(dihydroxyboranyl)phenyl]boronic acid
Synonyms: 4-(dihydroxyboranyl)phenylboronic acid
Molecular Weight (g/mol): 165.75
Molecular Formula: C6H8B2O4
InChi Key: BODYVHJTUHHINQ-UHFFFAOYSA-N
SMILES: OB(O)C1=CC=C(C=C1)B(O)O
Quantity 
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results