CAS RN 468740-43-4
CAS RN 468740-43-4
IUPAC Name:
4-{[2-(3-chlorophenyl)-2-hydroxyethyl]amino}-3-[4-methyl-6-(morpholin-4-yl)-2,3-dihydro-1H-1,3-benzodiazol-2-ylidene]-2,3-dihydropyridin-2-one
Synonyms:
4-{[2-(3-chlorophenyl)-2-hydroxyethyl]amino}-3-[4-methyl-6-(morpholin-4-yl)-1,3-dihydro-1,3-benzodiazol-2-ylidene]pyridin-2-one
Molecular Weight (g/mol):
479.97
Molecular Formula:
C25H26ClN5O3
InChi Key:
UGQMURPIMYALPH-UHFFFAOYNA-N
SMILES:
CC1=CC(=CC2=C1NC(N2)=C1C(=O)N=CC=C1NCC(O)C1=CC(Cl)=CC=C1)N1CCOCC1
1
–
2
of
2
results
1
Tocris Bioscience™ BMS 536924
CAS: 468740-43-4 Molecular Formula: C25H26ClN5O3
2
BMS-536924, MedChemExpress
MedChemExpress BMS-536924 is an orally active, competitive and selective insulin-like growth factor receptor (IGF-1R) kinase and insulin receptor (IR) inhibitor with IC50s of 100 nM and 73 nM, respectively. BMS-536924 has anti-cancer activity.