CAS RN 4692-98-2
IUPAC Name:
6-bromo-2,4-dihydro-1H-3,1-benzoxazine-2,4-dione
Synonyms:
6-bromo-1H-3,1-benzoxazine-2,4-dione
Molecular Weight (g/mol):
242.03
Molecular Formula:
C8H4BrNO3
InChi Key:
DXSMYDSFWCOSFM-UHFFFAOYSA-N
SMILES:
BrC1=CC=C2NC(=O)OC(=O)C2=C1
1
–
1
of
1
results
1
5-Bromoisatoic anhydride, 97%, Thermo Scientific Chemicals
CAS: 4692-98-2 Molecular Formula: C8H4BrNO3 Molecular Weight (g/mol): 242.03 MDL Number: MFCD00016921 InChI Key: DXSMYDSFWCOSFM-UHFFFAOYSA-N Synonym: 5-bromoisatoic anhydride,6-bromo-2h-3,1-benzoxazine-2,4 1h-dione,6-bromo-1h-benzo d 1,3 oxazine-2,4-dione,6-bromo isatinic anhydride,5-bromoisatoicanhydride,6-bromo-2,4-dihydro-1h-3,1-benzoxazine-2,4-dione,2h-3,1-benzoxazine-2,4 1h-dione, 6-bromo,6-bromo-1,2-dihydro-4h-3,1-benzoxazine-2,4-dione,5-bromo-isatoic anhydride,6-bromo-1h-benzo d 1,3-oxazaperhydroine-2,4-dione PubChem CID: 288561 IUPAC Name: 6-bromo-1H-3,1-benzoxazine-2,4-dione SMILES: BrC1=CC=C2NC(=O)OC(=O)C2=C1