CAS RN 4698-39-9
CAS RN 4698-39-9
IUPAC Name:
—
Synonyms:
—
Molecular Weight (g/mol):
—
Molecular Formula:
—
InChi Key:
—
SMILES:
—
1
–
1
of
1
results
1
(E)-Doxepin Hydrochloride, TRC
CAS: 4698-39-9 Molecular Formula: C19H22ClNO Molecular Weight (g/mol): 315.84 Synonym: (3E)-3-Dibenz[b,e]oxepin-11(6H)-ylidene-N,N-dimethyl-1-propanamine Hydrochloride,(E)-3-Dibenz[b,e]oxepin-11(6H)-ylidene-N,N-dimethyl-1-propanamine Hydrochloride,(E)-N,N-dimethyl-Dibenz[b,e]oxepin-delta11(6H),gamma-propylamine Hydrochloride,Dibenz[b,e]oxepin, 1-propanamine deriv.,(E)-Doxepin Hydrochloride,Doxepin Hydrochloride E isomer,trans-Doxepin Hydrochloride SMILES: Cl.CN(C)CC\C=C\1/c2ccccc2COc3ccccc13