CAS RN 470-17-7
CAS RN 470-17-7
IUPAC Name:
—
Synonyms:
—
Molecular Weight (g/mol):
—
Molecular Formula:
—
InChi Key:
—
SMILES:
—
1
–
1
of
1
results
1
Isoalantolactone, MedChemExpress
MedChemExpress Isoalantolactone is an apoptosis inducer, which also acts as an alkylating agent.