accessibility menu, dialog, popup

UserName

H3COPOCH3OCH3

CAS RN 4731-65-1

IUPAC Name: tris(2-methoxyphenyl)phosphane
Synonyms: tris(2-methoxyphenyl)phosphane
Molecular Weight (g/mol): 352.37
Molecular Formula: C21H21O3P
InChi Key: IIOSDXGZLBPOHD-UHFFFAOYSA-N
SMILES: COC1=CC=CC=C1P(C1=CC=CC=C1OC)C1=CC=CC=C1OC
Percent Purity 
  • (2)
  • (3)
Quantity 
  • (2)
  • (1)
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results