CAS RN 4732-82-5
CAS RN 4732-82-5
IUPAC Name:
—
Synonyms:
—
Molecular Weight (g/mol):
—
Molecular Formula:
—
InChi Key:
—
SMILES:
—
1
–
1
of
1
results
1
3’-Iodo-L-thyronine, TRC
CAS: 4732-82-5 Molecular Formula: C15H14INO4 Molecular Weight (g/mol): 399.18 Synonym: 3-[p-(4-Hydroxy-3-iodophenoxy)phenyl]-alanine,3'-Iodothyronine SMILES: IC1=C(O)C=CC(OC2=CC=C(C[C@H](N)C(O)=O)C=C2)=C1