accessibility menu, dialog, popup

UserName

H3CCH3H3COONBrBHOHO

CAS RN 475102-13-7

IUPAC Name: {5-bromo-1-[(tert-butoxy)carbonyl]-1H-indol-2-yl}boronic acid
Synonyms: 5-bromo-1-(tert-butoxycarbonyl)indol-2-ylboronic acid
Molecular Weight (g/mol): 339.98
Molecular Formula: C13H15BBrNO4
InChi Key: RBYTXZMVOGZESQ-UHFFFAOYSA-N
SMILES: CC(C)(C)OC(=O)N1C(=CC2=CC(Br)=CC=C12)B(O)O

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results