accessibility menu, dialog, popup

UserName

H3COOHNCl

CAS RN 4792-67-0

IUPAC Name: ethyl 5-chloro-1H-indole-2-carboxylate
Synonyms: ethyl 5-chloro-1H-indole-2-carboxylate
Molecular Weight (g/mol): 223.66
Molecular Formula: C11H10ClNO2
InChi Key: LWKIFKYHCJAIAB-UHFFFAOYSA-N
SMILES: CCOC(=O)C1=CC2=CC(Cl)=CC=C2N1
Molecular Weight (g/mol) 
  • (3)
  • (2)
Percent Purity 
  • (2)
  • (3)
Quantity 
  • (1)
  • (1)
  • (1)
  • (2)
Physical Form 
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (3)
  • (3)
  • (2)
  • (2)
  • (3)
  • (1)
  • (1)
  • (1)
  • (2)
  • (2)