accessibility menu, dialog, popup

UserName

CAS RN 4795-29-3

H2NO

CAS RN 4795-29-3

IUPAC Name: 1-(oxolan-2-yl)methanamine
Synonyms: 1-(oxolan-2-yl)methanamine
Molecular Weight (g/mol): 101.15
Molecular Formula: C5H11NO
InChi Key: YNOGYQAEJGADFJ-UHFFFAOYNA-N
SMILES: NCC1CCCO1
Boiling Point 
  • (2)
  • (3)
Quantity 
  • (2)
  • (1)
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (1)
  • (3)
  • (2)
  • (2)
  • (3)
  • (2)
  • (1)
  • (2)