CAS RN 481-85-6
CAS RN 481-85-6
IUPAC Name:
—
Synonyms:
—
Molecular Weight (g/mol):
—
Molecular Formula:
—
InChi Key:
—
SMILES:
—
1
–
1
of
1
results
1
2-Methyl-1,4-naphthalenediol, TRC
CAS: 481-85-6 Molecular Formula: C11 H10 O2 Molecular Weight (g/mol): 174.2 Synonym: 1,4-Dihydroxy-2-methylnaphthalene,2-Methyl-1,4-dihydroxynaphthalene,2-Methyl-1,4-naphthohydroquinone,2-Methyl-1,4-naphthoquinol,2-Methylhydronaphthoquinone,2-Methylnaphthalene-1,4-diol,4-Hydroxy-3-methyl-α-naphthol,Dihydrovitamin K3,Menadiol,Reduced Menadione,Reduced Vitamin K3,Vitamin K3H2 IUPAC Name: 2-methylnaphthalene-1,4-diol SMILES: Cc1cc(O)c2ccccc2c1O