CAS RN 486-79-3
CAS RN 486-79-3
IUPAC Name:
—
Synonyms:
—
Molecular Weight (g/mol):
—
Molecular Formula:
—
InChi Key:
—
SMILES:
—
1
–
1
of
1
results
1
Dipyrocetyl, MedChemExpress
MedChemExpress Dipyrocetyl is an anti-inflammatory and analgesic agent extracted from patent WO 2011132171 A1.