accessibility menu, dialog, popup

UserName

CAS RN 487-60-5

HO(R)O(S)OHN(R)OH(S)HO(S)HO

CAS RN 487-60-5

IUPAC Name: (2R,3S,4S,5R,6S)-2-(hydroxymethyl)-6-(1H-indol-3-yloxy)oxane-3,4,5-triol
Synonyms: indican plant indican indoxyl-β-D-glucoside indikan indican (glucoside) indican, plant 3-indoxylsulfuric acid uroxanthin 3-(glucosyloxy)indole indoxyl sulfic acid
Molecular Weight (g/mol): 295.29
Molecular Formula: C14H17NO6
InChi Key: XVARCVCWNFACQC-RKQHYHRCSA-N
SMILES: OC[C@H]1O[C@@H](OC2=CNC3=CC=CC=C23)[C@H](O)[C@@H](O)[C@@H]1O

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (1)
  • (2)
  • (2)
  • (1)
  • (1)