accessibility menu, dialog, popup

UserName

CAS RN 487051-12-7

IUPAC Name: N-[3-(1-{[4-(3,4-difluorophenoxy)phenyl]methyl}piperidin-4-yl)-4-methylphenyl]-2-methylpropanamide hydrochloride
Synonyms: N-[3-(1-{[4-(3,4-difluorophenoxy)phenyl]methyl}piperidin-4-yl)-4-methylphenyl]-2-methylpropanamide hydrochloride
Molecular Weight (g/mol): 515.04
Molecular Formula: C29H33ClF2N2O2
InChi Key: DEDUDFNRQKUBRH-UHFFFAOYSA-N
SMILES: Cl.CC(C)C(=O)NC1=CC(C2CCN(CC3=CC=C(OC4=CC(F)=C(F)C=C4)C=C3)CC2)=C(C)C=C1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results