accessibility menu, dialog, popup

UserName

H3CCH3OSOOCH3

CAS RN 4873-56-7

IUPAC Name: 2-methylpropyl 4-methylbenzene-1-sulfonate
Synonyms: 2-methylpropyl 4-methylbenzenesulfonate
Molecular Weight (g/mol): 228.31
Molecular Formula: C11H16O3S
InChi Key: QIMRPGAQCKHRKB-UHFFFAOYSA-N
SMILES: CC(C)COS(=O)(=O)C1=CC=C(C)C=C1
Molecular Weight (g/mol) 
  • (2)
  • (3)
Quantity 
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (3)
  • (2)
  • (3)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)