CAS RN 4921-49-7
IUPAC Name:
—
Synonyms:
—
Molecular Weight (g/mol):
—
Molecular Formula:
—
InChi Key:
—
SMILES:
—
1
–
2
of
2
results
1
8-Chloro-1,3,7-trimethyl-3,7-dihydro-1H-purine-2,6-dione (8-Chlorocaffeine), Mikromol™
Discover Mikromol - your trusted source for high-quality pharmaceutical reference standards. Supporting accurate, compliant analysis with ISO 17034-accredited materials designed for confidence in every result.
2
8-Chloro Caffeine, TRC
CAS: 4921-49-7 Molecular Formula: C8 H9 Cl N4 O2 Molecular Weight (g/mol): 228.64 Synonym: 8-Chloro-1,3,7-trimethyl-3,7-dihydro-1H-purine-2,6-dione,Dimenhydrinate Imp. E (EP),8-Chlorocaffeine IUPAC Name: 8-chloro-1,3,7-trimethylpurine-2,6-dione SMILES: CN1C(=O)N(C)c2nc(Cl)n(C)c2C1=O