CAS RN 4930-98-7
CAS RN 4930-98-7
IUPAC Name:
2-hydrazinylpyridine
Synonyms:
2(1H)-pyridinone, hydrazone
Molecular Weight (g/mol):
109.13
Molecular Formula:
C5H7N3
InChi Key:
NWELCUKYUCBVKK-UHFFFAOYSA-N
SMILES:
NNC1=CC=CC=N1
No Refinements
1
–
0
of
0
results
1
2-Hydrazinopyridine, 98%
CAS: 4930-98-7 Molecular Formula: C5H7N3 Molecular Weight (g/mol): 109.13 MDL Number: MFCD00006249 InChI Key: NWELCUKYUCBVKK-UHFFFAOYSA-N Synonym: 2-hydrazinylpyridine,2-hydrazinopyridine,2-pyridylhydrazine,2 1h-pyridinone, hydrazone,hydrazine, 2-pyridinyl,pyridine, 2-hydrazinyl,2-pyridylhydrazone,pyridin-2-yl-hydrazine,pyridine, 2-hydrazino,pyridin-2-yl hydrazine PubChem CID: 78645 IUPAC Name: pyridin-2-ylhydrazine SMILES: NNC1=CC=CC=N1