accessibility menu, dialog, popup

UserName

CAS RN 496-14-0

IUPAC Name: 1,3-dihydro-2-benzofuran
Synonyms: phthalan phthalane
Molecular Weight (g/mol): 120.15
Molecular Formula: C8H8O
InChi Key: SFLGSKRGOWRGBR-UHFFFAOYSA-N
SMILES: C1OCC2=CC=CC=C12
Percent Purity 
  • (1)
  • (3)
Quantity 
  • (1)
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results