CAS RN 49691-65-8
IUPAC Name:
—
Synonyms:
—
Molecular Weight (g/mol):
—
Molecular Formula:
—
InChi Key:
—
SMILES:
—
1
–
1
of
1
results
1
1-Chloroacetyl-2-[[(2-benzoyl-4-chlorophenyl)(2-chloroacetyl)amino]methylene]hydrazide, TRC
CAS: 49691-65-8 Molecular Formula: C18 H14 Cl3 N3 O3 Molecular Weight (g/mol): 426.68 Synonym: 2-Chloroacetic Acid 2-[[(2-Benzoyl-4-chlorophenyl)(2-chloroacetyl)amino]methylene]hydrazide,Chloroacetic Acid [[(2-Benzoyl-4-chlorophenyl)(chloroacetyl)amino]methylene]hydrazide,(E)-N-(2-Benzoyl-4-chlorophenyl)-2-chloro-N-((2-(2-chloroacetyl)hydrazono)methyl)acetamide,1-Chloroacetyl-2-[[(2-benzoyl-4-chlorophenyl)(2-chloroacetyl)amino]methylene]hydrazide IUPAC Name: N-[(E)-(2-benzoyl-4-chloro-N-(2-chloroacetyl)anilino)methylideneamino]-2-chloroacetamide SMILES: ClCC(=O)N\N=C\N(C(=O)CCl)c1ccc(Cl)cc1C(=O)c2ccccc2