accessibility menu, dialog, popup

UserName

CAS RN 500-73-2

FFFOO

CAS RN 500-73-2

IUPAC Name: phenyl 2,2,2-trifluoroacetate
Synonyms: phenyl 2,2,2-trifluoroacetate
Molecular Weight (g/mol): 190.12
Molecular Formula: C8H5F3O2
InChi Key: DVCMYAIUSOSIQP-UHFFFAOYSA-N
SMILES: FC(F)(F)C(=O)OC1=CC=CC=C1
Boiling Point 
  • (2)
  • (1)
Molecular Weight (g/mol) 
  • (1)
  • (2)
Quantity 
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (1)
  • (2)
  • (2)
  • (1)
  • (1)
  • (2)
  • (1)
  • (1)
  • (1)
  • (1)