CAS RN 500536-41-4
IUPAC Name:
2,7-dimethyl-9-oxo-9H-fluorene-4-carboxylic acid
Synonyms:
2,7-dimethyl-9-oxofluorene-4-carboxylic acid
Molecular Weight (g/mol):
252.27
Molecular Formula:
C16H12O3
InChi Key:
ATPRNYZONXAEOW-UHFFFAOYSA-N
SMILES:
CC1=CC2=C(C=C1)C1=C(C=C(C)C=C1C2=O)C(O)=O
1
–
1
of
1
results
1
2,7-Dimethyl-9-fluorenone-4-carboxylic acid, 97%, Thermo Scientific™
CAS: 500536-41-4 Molecular Formula: C16H12O3 Molecular Weight (g/mol): 252.269 MDL Number: MFCD04039780 InChI Key: ATPRNYZONXAEOW-UHFFFAOYSA-N Synonym: 2,7-dimethyl-9-oxo-9h-fluorene-4-carboxylic acid,2,7-dimethyl-9-fluorenone-4-carboxylic acid PubChem CID: 246697 IUPAC Name: 2,7-dimethyl-9-oxofluorene-4-carboxylic acid SMILES: CC1=CC2=C(C=C1)C3=C(C=C(C=C3C2=O)C)C(=O)O