CAS RN 501697-80-9
IUPAC Name:
3',4'-dichloro-[1,1'-biphenyl]-4-sulfonyl chloride
Synonyms:
3',4'-dichloro-[1,1'-biphenyl]-4-sulfonyl chloride
Molecular Weight (g/mol):
321.6
Molecular Formula:
C12H7Cl3O2S
InChi Key:
DHVBZUPMGVOIOH-UHFFFAOYSA-N
SMILES:
ClC1=C(Cl)C=C(C=C1)C1=CC=C(C=C1)S(Cl)(=O)=O
1
–
1
of
1
results
1
3',4'-Dichlorobiphenyl-4-sulfonyl chloride, 95%, Thermo Scientific™
CAS: 501697-80-9 Molecular Formula: C12H7Cl3O2S Molecular Weight (g/mol): 321.61 InChI Key: DHVBZUPMGVOIOH-UHFFFAOYSA-N Synonym: 3',4'-dichloro 1,1'-biphenyl-4-sulfonyl chloride,4-3,4-dichlorophenyl benzenesulfonyl chloride,3',4'-dichloro-biphenyl-4-sulfonyl chloride,3',4'-dichloro-1,1'-biphenyl-4-sulfonyl chloride,4-3,4-dichlorophenyl phenyl chlorosulfone,4-3,4-dichlorophenyl phenyl sulfonyl chloride,4-3,4-dichlorophenyl phenyl sulphonyl chloride PubChem CID: 2794751 IUPAC Name: 4-(3,4-dichlorophenyl)benzenesulfonyl chloride SMILES: C1=CC(=CC=C1C2=CC(=C(C=C2)Cl)Cl)S(=O)(=O)Cl