CAS RN 5018-38-2
IUPAC Name:
4,6-dichloro-5-methoxypyrimidine
Synonyms:
4,6-dichloro-5-methoxypyrimidine
Molecular Weight (g/mol):
179
Molecular Formula:
C5H4Cl2N2O
InChi Key:
IJQIGKLDBGKSNT-UHFFFAOYSA-N
SMILES:
COC1=C(Cl)N=CN=C1Cl
1
–
1
of
1
results
1
4,6-Dichloro-5-methoxypyrimidine, 97%, Thermo Scientific™
CAS: 5018-38-2 Molecular Formula: C5H4Cl2N2O Molecular Weight (g/mol): 179 MDL Number: MFCD00087680 InChI Key: IJQIGKLDBGKSNT-UHFFFAOYSA-N Synonym: 4,6-dichlor-5-methoxypyrimidin,4,6-dichloro-5-methoxy-pyrimidine,4,6-dichloro-5-methoxy pyrimidine,zlchem 448,pubchem7077,pyrimidine, 4,6-dichloro-5-methoxy,acmc-1avm3,4,6 dichloro-5-methoxypyrimidine,pyrimidine,6-dichloro-5-methoxy,4,6-dichloro-5-methoxypyrimidine PubChem CID: 78720 IUPAC Name: 4,6-dichloro-5-methoxypyrimidine SMILES: COC1=C(N=CN=C1Cl)Cl